missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLG1 Antibody (RPI 197.4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-88994
This item is not returnable.
View return policy
Description
DLG1 Monoclonal specifically detects DLG1 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Electron Microscopy.
Specifications
| DLG1 | |
| Monoclonal | |
| Unconjugated | |
| discs, large homolog 1 (Drosophila), disks large homolog 1, DKFZp761P0818, DKFZp781B0426, hDlg, large (Drosophila) homolog 1, presynaptic protein SAP97 | |
| Recombinant rat DLG1(N-terminal fragment). | |
| 200 ug | |
| Neuroscience, Vision | |
| 1739 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunomicroscopy | |
| RPI 197.4 | |
| PBS, pH 7.2, containing 50% glycerol | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction