missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLG1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | DLG1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Applications | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30228803
|
Novus Biologicals
NBP3-38560-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30231761
|
Novus Biologicals
NBP3-38560-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DLG1 Polyclonal antibody specifically detects DLG1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ImmunofluorescenceSpecifications
| DLG1 | |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Neuroscience, Vision | |
| PBS (pH 7.3), 50% glycerol | |
| 1739 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| discs, large homolog 1 (Drosophila), disks large homolog 1, DKFZp761P0818, DKFZp781B0426, hDlg, large (Drosophila) homolog 1, presynaptic protein SAP97 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human DLG1 (NP_004078.2).,, Sequence:, MPVRKQDTQRALHLLEEYRSKLSQTEDRQLRSSIERVINIFQSNLFQALIDIQEFYEVTLLDNPKCIDRSKPSEPIQPVNTWEISSLPSSTVTSETLPSSLSPSVEKYRYQDEDTPPQEHISPQITNEVIGPELVHVSEKNLSEIENVHGFVSHSHISPIKPTEAVLPSPPTVPVIPVLPVPAENTVILP | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title