missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLG1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-26611
This item is not returnable.
View return policy
Description
DLG1 Polyclonal specifically detects DLG1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| DLG1 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| discs, large homolog 1 (Drosophila), disks large homolog 1, DKFZp761P0818, DKFZp781B0426, hDlg, large (Drosophila) homolog 1, presynaptic protein SAP97 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 1739 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 5-10 ug/mg lysate | |
| Q12959 | |
| DLG1 | |
| The immunogen recognized by this antibody maps to a region between residue 50 and 100 of human synapse-associated protein 97 (discs, large homolog 1) using the numbering given in entry NP_004078.1 (GeneID 1739 ). | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction