missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DLEC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553917
This item is not returnable.
View return policy
Description
Immunogen sequence: PPVKSVSRWC IDSELLRKHH LISPEDYYTD TVPFHSAPKG ISLPGCSKLT FSCEKRSVQK KELNKKLEDS CRKKLAEFED ELDHTVDSLT WNLTPKAKER TREPLKKASQ PRN Highest antigen sequence identity to the following orthologs: Mouse - 58%, Rat - 56%.
This gene contains 37 exons, spans approximately 59-kb, and is located in the 3p22-p21.3 chromosomal segment that is commonly deleted in various carcinomas. Several alternatively spliced transcripts have been observed that contain disrupted coding regions and likely encode nonfunctional proteins. Aberrant transcription of this gene may be involved in carcinogenesis of the lung, esophagus, and kidney.
Specifications
| DLEC1 | |
| Polyclonal | |
| Unconjugated | |
| DLEC1 | |
| CFAP81; cilia and flagella associated protein 81; D630005C06Rik; deleted in lung and esophageal cancer 1; deleted in lung and esophageal cancer protein 1; deleted in lung and esophageal cancer protein 1 homolog; Deleted in lung cancer protein 1; DLC1; DLC-1; DLEC1; F56 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9940 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.6 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y238 | |
| DLEC1 | |
| Recombinant protein corresponding to Human DLEC1. Recombinant protein control fragment (Product #RP-92502). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction