missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ DLEC/CLEC4C/BDCA-2 Antibody (LFB patent anti-BDCA-2) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
£162.00 - £695.00
Specifications
| Antigen | DLEC/CLEC4C/BDCA-2 |
|---|---|
| Clone | LFB patent anti-BDCA-2 |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30046900
|
Novus Biologicals™
NBP3-28633-100ug |
100 μg |
£257.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30046364
|
Novus Biologicals™
NBP3-28633-50ug |
50 μg |
£162.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30046351
|
Novus Biologicals™
NBP3-28633-1mg |
1 mg |
£695.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DLEC/CLEC4C/BDCA-2 Monoclonal antibody specifically detects DLEC/CLEC4C/BDCA-2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| DLEC/CLEC4C/BDCA-2 | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Dendritic Cell Markers | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| BDCA2, BDCA-2, BDCA2MGC125791, Blood dendritic cell antigen 2, blood dendritic cell antigen 2 protein, CD303, CD303 antigen, CLECSF11, CLECSF11PRO34150, CLECSF7, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 11, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 7, C-type lectin domain family 4 member C, C-type lectin superfamily member 7, dendritic cell lectin b, Dendritic lectin, DLEC, DLECMGC125792, HECL, HECLMGC125793, MGC125789, PRO34150 | |
| CLEC4C | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| LFB patent anti-BDCA-2 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q8WTT0 | |
| 170482 | |
| IgG | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title