missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DLD Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5122035
This item is not returnable.
View return policy
Description
DLD Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes the L protein of the mitochondrial glycine cleavage system. The L protein, also named dihydrolipoamide dehydrogenase, is also a component of the pyruvate dehydrogenase complex, the alpha-ketoglutarate dehydrogenase complex, and the branched-chain alpha-keto acide dehydrogenase complex. Mutations in this gene have been identified in patients with E3-deficient maple syrup urine disease and lipoamide dehydrogenase deficiency.
Specifications
| DLD | |
| Polyclonal | |
| Unconjugated | |
| Dld | |
| aei; after eight protein; branched chain alpha-keto acid dehydrogenase complex subunit E3; deltaD; deltaD transmembrane protein; delta-like protein D; diaphorase; dihydrolipoamide dehydrogenase; dihydrolipoamide dehydrogenase (E3 component of pyruvate dehydrogenase complex, 2-oxo-glutarate complex, branched chain keto acid dehydrogenase complex); dihydrolipoamide: NAD+ oxidoreductase; dihydrolipoyl dehydrogenase mitochondrial-like protein; dihydrolipoyl dehydrogenase, mitochondrial; DLD; DLDD; DLDH; E3; E3 component of pyruvate dehydrogenase complex, 2-oxo-glutarate complex, branched chain keto acid dehydrogenase complex; GCSL; Glycine cleavage system L protein; glycine cleavage system protein L; LAD; lipoamide dehydrogenase; lipoamide dehydrogenase precursor; lipoamide reductase; lipoyl dehydrogenase; LOW QUALITY PROTEIN: dihydrolipoyl dehydrogenase, mitochondrial; mitochondrial Dihydrolipoyl dehydrogenase-like protein; OTTHUMP00000206744; OTTHUMP00000206746; OTTHUMP00000206748; OTTHUMP00000206749; PH; PHE 3; PHE3; QflA-16416; QtsA-11266; sc:d0402; unnamed protein product | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 13382, 1738 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O08749, P09622 | |
| Dld | |
| Recombinant protein Dihydrolipoyl Dehydrogenase. The antigen corresponds to amino acid range 280-487 of the target protein. | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction