missing translation for 'onlineSavingsMsg'
Learn More

DLD Mouse Monoclonal Antibody, Proteintech

Mouse Monoclonal Antibody

£119.00 - £295.00

Specifications

Antigen DLD
Clone 2E7G5
Concentration 1100 μg/mL
Applications Western Blot, Immunohistochemistry, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30233390
View Documents
Proteintech
67702-1-IG-150UL
150 μL
ONLINE EXCLUSIVE PRICE
£349.00
£295.00 / 150µL
Save £54.00
15% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30234329
View Documents
Proteintech
67702-1-IG-20UL
20 μL
ONLINE EXCLUSIVE PRICE
£135.00
£119.00 / 20µL
Save £16.00
11% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The DLD antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DLD. This antibody recognizes human, mouse, rat antigen. The DLD antibody has been validated for the following applications: WB, IHC, ELISA analysis.
Specifications

Specifications

DLD
1100 μg/mL
Monoclonal
Liquid
RUO
1738
Recombinant Protein
Primary
Store at -20°C.
2E7G5
Western Blot, Immunohistochemistry, ELISA
Unconjugated
Mouse
Human, Mouse, Rat
DLDH; E3; GCSL; LAD; PHE3
IgG1
Protein G purification
Monoclonal Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.