missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15460-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a GTPase-activating protein that is a member of the rhoGAP family of proteins which play a role in the regulation of small GTP-binding proteins. GAP family proteins participate in signaling pathways that regulate cell processes involved in cytoskeletal changes. This gene functions as a tumor suppressor gene in a number of common cancers, including prostate, lung, colorectal, and breast cancers. Multiple transcript variants due to alternative promoters and alternative splicing have been found for this gene.Specifications
| DLC1 | |
| Polyclonal | |
| Unconjugated | |
| DLC1 | |
| ARHGAP7, deleted in liver cancer 1, DLC 1, DLC1, HP, HP protein, KIAA1723, p122 RhoGAP, STARD12 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10395, 50768, 58834 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q63744, Q96QB1, Q9R0Z9 | |
| Dlc1 | |
| DLC1 Fusion Protein Ag7732 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction