missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DLC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569821
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 79%.
This gene encodes a GTPase-activating protein that is a member of the rhoGAP family of proteins which play a role in the regulation of small GTP-binding proteins. GAP family proteins participate in signaling pathways that regulate cell processes involved in cytoskeletal changes. This gene functions as a tumor suppressor gene in a number of common cancers, including prostate, lung, colorectal, and breast cancers. Multiple transcript variants due to alternative promoters and alternative splicing have been found for this gene.
Specifications
| DLC1 | |
| Polyclonal | |
| Unconjugated | |
| Dlc1 | |
| A730069N07Rik; ARHGAP 7; ARHGAP7; deleted in liver cancer 1; deleted in liver cancer 1 protein; Deleted in liver cancer 1 protein homolog; deleted in liver cancer 1 variant 2; deleted in liver cancer variant 4; DLC 1; DLC1; Dlc-1; DLC1 Rho GTPase activating protein; DLC1 Rho GTPase activating protein; rho GTPase-activating protein 7; FLJ21120; HP; HP protein; KIAA1723; p122 RhoGAP; p122-RhoGAP; rho GTPase activating protein 7; rho GTPase-activating protein 7; RhoGAP; Rho-GTPase-activating protein 7; rho-type GTPase-activating protein 7; STARD 12; STARD12; StAR-related lipid; StAR-related lipid transfer (START) domain containing 12; stAR-related lipid transfer protein 12; START domain-containing protein 12; transfer protein 12; unm_sa1141 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10395 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96QB1 | |
| Dlc1 | |
| synthetic peptide directed towards the C terminal region of human DLC1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction