missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DLC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106902
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total DLC1.
This gene encodes a GTPase-activating protein that is a member of the rhoGAP family of proteins which play a role in the regulation of small GTP-binding proteins. GAP family proteins participate in signaling pathways that regulate cell processes involved in cytoskeletal changes. This gene functions as a tumor suppressor gene in a number of common cancers, including prostate, lung, colorectal, and breast cancers. Multiple transcript variants due to alternative promoters and alternative splicing have been found for this gene.
Specifications
| DLC1 | |
| Polyclonal | |
| Unconjugated | |
| Dlc1 | |
| A730069N07Rik; ARHGAP 7; ARHGAP7; deleted in liver cancer 1; deleted in liver cancer 1 protein; Deleted in liver cancer 1 protein homolog; deleted in liver cancer 1 variant 2; deleted in liver cancer variant 4; DLC 1; DLC1; Dlc-1; DLC1 Rho GTPase activating protein; DLC1 Rho GTPase activating protein; rho GTPase-activating protein 7; FLJ21120; HP; HP protein; KIAA1723; p122 RhoGAP; p122-RhoGAP; rho GTPase activating protein 7; rho GTPase-activating protein 7; RhoGAP; Rho-GTPase-activating protein 7; rho-type GTPase-activating protein 7; STARD 12; STARD12; StAR-related lipid; StAR-related lipid transfer (START) domain containing 12; stAR-related lipid transfer protein 12; START domain-containing protein 12; transfer protein 12; unm_sa1141 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10395, 50768, 58834 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q63744, Q96QB1, Q9R0Z9 | |
| Dlc1 | |
| A synthesized peptide derived from human DLC1(Accession Q96QB1), corresponding to amino acid residues N51-E101. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction