missing translation for 'onlineSavingsMsg'
Learn More

DLC1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£145.00 - £417.00

Specifications

Antigen DLC1
Dilution Western Blot 1:500 - 1:2000, ELISA
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30229637
View Documents
Novus Biologicals
NBP3-35894-20ul
20 μL
£145.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30229843
View Documents
Novus Biologicals
NBP3-35894-100ul
100 μL
£417.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DLC1 Polyclonal antibody specifically detects DLC1 in Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications

Specifications

DLC1
ELISA, Western Blot
Unconjugated
Rabbit
Mouse, Rat
ARHGAP7 STARD12 deleted in liver cancer 1 variant 2, deleted in liver cancer 1, Deleted in liver cancer 1 protein, DLC-1, FLJ21120, HP, HP protein, KIAA1723, p122-RhoGAP, rho GTPase-activating protein 7, Rho-GTPase-activating protein 7, Rho-type GTPase-activating protein 7, StARD12, StAR-related lipid transfer (START) domain containing 12, StAR-related lipid transfer protein 12, START domain-containing protein 12
Recombinant fusion protein containing a sequence corresponding to amino acids 204-463 of human DLC1 (NP_079043.3).,, Sequence:, DAPKVNAVDTLNVKDIAPEKQLLNSAVIAQQRRKPDPPKDENERSTCNVVQNEFLDTPCTNRGLPLLKTDFGSCLLQPPSCPNGMSAENGLEKSGFSQHQNKSPPKVKAEDGMQCLQLKETLATQEPTDNQVRLRKRKEIREDRDRARLDSMVLLIMKLDQLDQDIENALSTSSSPSGTPTNLRRHVPDLESGSESGADTISVNQTRVNLSSDTESTDLPSSTPVANSGTKPKTTAIQGISEKEKAGKLTFWFCFLANLF
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot 1:500 - 1:2000, ELISA
Polyclonal
Purified
RUO
PBS (pH 7.3), 50% glycerol
10395
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.