missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DLC1 Polyclonal antibody specifically detects DLC1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | DLC1 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.04 - 0.4 ug/mL, Immunohistochemistry 1:2500 - 1:5000, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:2500-1:5000 |
| Formulation | PBS (pH 7.2) and 40% Glycerol |
| Gene Alias | ARHGAP7 STARD12 deleted in liver cancer 1 variant 2, deleted in liver cancer 1, Deleted in liver cancer 1 protein, DLC-1, FLJ21120, HP, HP protein, KIAA1723, p122-RhoGAP, rho GTPase-activating protein 7, Rho-GTPase-activating protein 7, Rho-type GTPase-activating protein 7, StARD12, StAR-related lipid transfer (START) domain containing 12, StAR-related lipid transfer protein 12, START domain-containing protein 12 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: RSWEEHVTHWMGQPFNSDDRNTACHHGLVADSLQASMEKDATLNVDRKEKCVSLPDCCHGSELRDFPGRPMGHLSKDVDENDSHEGEDQFLSLEASTETLVHVSDEDNNADLCLTDDKQVLNTQGQ |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?