Learn More
Invitrogen™ DLAT Monoclonal Antibody (K29029_13C6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555988
Description
Sequence of this protein is as follows: HQKVPLPSLS PTMQAGTIAR WEKKEGDKIN EGDLIAEVET DKATVGFESL EECYMAKILV AEGTRDVPIG AIICITVGKP EDIEAFKNYT LDSSAAPTPQ AAPAPTPAAT ASPPTPSAQA PGSSYPPHMQ VLLPALSPTM TMGTVQRWEK KVGEKLSEGD LLAEIETDKA TIGFEVQEEG YLAKILVPEG TRDVPLGTPL CIIVEKEADI SAFADYRPTE VTDLKPQVPP PTPPPVAAVP PTPQPLAPTP SAPCPATPAG PKGRVFVSPL AKKLAVEKGI DLTQVKGTGP DGRITKKDID SFVPSKVAPA PAAVVPPTGP
DLAT encodes component E2 of the multi-enzyme pyruvate dehydrogenase complex (PDC). PDC resides in the inner mitochondrial membrane and catalyzes the conversion of pyruvate to acetyl coenzyme A. The protein product of this gene, dihydrolipoamide acetyltransferase, accepts acetyl groups formed by the oxidative decarboxylation of pyruvate and transfers them to coenzyme A. Dihydrolipoamide acetyltransferase is the antigen for antimitochondrial antibodies. These autoantibodies are present in nearly 95% of patients with the autoimmune liver disease primary biliary cirrhosis (PBC). In PBC, activated T lymphocytes attack and destroy epithelial cells in the bile duct where this protein is abnormally distributed and overexpressed. PBC eventually leads to cirrhosis and liver failure.
Specifications
| DLAT | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| P10515 | |
| DLAT | |
| Recombinant human DLAT (immunogen range is 91-410). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| K29029_13C6 | |
| Unconjugated | |
| DLAT | |
| 6332404G05Rik; 70 kDa mitochondrial autoantigen of primary biliary cirrhosis; anti DLAT; Dihydrolipoamide; Dihydrolipoamide acetyltransferase component of pyruvate dehydrogenase complex; dihydrolipoamide S-acetyltransferase; dihydrolipoamide S-acetyltransferase (E2 component of pyruvate dehydrogenase complex); dihydrolipoyllysine-residue acetyltransferase component of pyruvate dehydrogenase complex mitochondrial; dihydrolipoyllysine-residue acetyltransferase component of pyruvate dehydrogenase complex, mitochondrial; DLAT; DLTA; E2; E2 component of pyruvate dehydrogenase complex; EC 2.3.1.12; M2 antigen complex 70 kDa subunit; ODP2; PBC; PDC PDC-E2; PDCE2; PDC-E2; pyruvate dehydrogenase complex component E2 | |
| Mouse | |
| Protein A | |
| RUO | |
| 1737 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.