missing translation for 'onlineSavingsMsg'
Learn More

DL-Tryptophan-d8, CDN

Product Code. 30319520
Change view
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Product Code. Quantity unitSize
30319520 0.05 g 0.05g
30310639 0.01 g 0.01g
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30319520

Brand: CDN CDND16480.05G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DL-Tryptophan-d8
CAS 1233395-85-1
Molecular Formula C11 D8 H4 N2 O2
Quantity 0.05 g
Synonym Tryptophan-α,β,β,2,4,5,6,7-d8 (ACI)
InChI Formula InChI=1 S/C11H12N2O2/c12-9(11(14)15)5-7-6-13-10-4-2-1-3-8(7)10/h1-4,6,9,13 H,5,12H2,(H,14,15)/i1D,2 D,3 D,4 D,5D2,6 D,9 D
SMILES [2 H]c1[nH]c2c([2 H])c([2 H])c([2 H])c([2 H])c2c1C([2 H])([2 H])C([2 H])(N)C(=O)O
IUPAC Name 2-amino-2,3,3-trideuterio-3-(2,4,5,6,7-pentadeuterio-1 H-indol-3-yl)propanoic acid
Molecular Weight (g/mol) 212.2745
Formula Weight 212.1401 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.