missing translation for 'onlineSavingsMsg'
Learn More

DL-Tropic acid, 97%

Product Code. p-3758128 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10530902 5 g Glass bottle 5g
10523691 25 g Glass bottle 25g
10579053 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10530902 Supplier Thermo Scientific Acros Supplier No. 140550050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 552-63-6 | C9H10O3 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 552-63-6
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00004255
InChI Key JACRWUWPXAESPB-UHFFFAOYNA-N
Synonym tropic acid, dl-tropic acid, 2-phenylhydracrylic acid, 3-hydroxy-2-phenylpropionic acid, +--tropic acid, tropate, alpha-hydroxymethyl benzeneacetic acid, beta-hydroxyhydratropic acid, hydracrylic acid, 2-phenyl, alpha-hydroxymethyl phenylacetic acid
PubChem CID 10726
ChEBI CHEBI:30765
IUPAC Name 3-hydroxy-2-phenylpropanoic acid
SMILES OCC(C(O)=O)C1=CC=CC=C1

Specifications

Melting Point 117.0°C to 120.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH(CH2OH)CO2H
Quantity 5 g
Beilstein 10, 261
Merck Index 15, 9958
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol and ether,slightly soluble in benzene,practically insoluble in petroleum ether
Formula Weight 166.18
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material DL-Tropic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.