missing translation for 'onlineSavingsMsg'
Learn More

DL-Threonic Acid Calcium, TRC

Product Code. 30456403
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456403 10 mg 10mg
30454733 1 mg 1mg
30455989 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456403 Supplier Toronto Research Chemicals Supplier No. TRCT40558810MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DL-Threonic Acid Calcium
Analyte or Component Names DL-Threonic Acid Calcium
Shipping Condition Room Temperature
Molecular Formula C8H14CaO10
Synonym calcium (2S,3R)-2,3,4-trihydroxybutanoate
InChI Formula InChI=1S/2C4H8O5.Ca/c2*5-1-2(6)3(7)4(8)9;/h2*2-3,5-7H,1H2,(H,8,9);/q;;+2/p-2/t2*2-,3+;/m11./s1
Solution Type Neat
SMILES O=C([C@H]([C@@H](CO)O)O)[O-].O=C([C@H]([C@@H](CO)O)O)[O-].[Ca+2]
IUPAC Name calcium (2S,3R)-2,3,4-trihydroxybutanoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 310.27
Quantity 10 mg
Formula Weight 310.02
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.