missing translation for 'onlineSavingsMsg'
Learn More

DL-Theanine, TRC

Product Code. 30452046
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
2.5mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30452046 25 mg 25mg
30455623 5 mg 5mg
30455591 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30452046 Supplier Toronto Research Chemicals Supplier No. TRCT40549825MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34271-54-0
Molecular Formula C7H14N2O3
Molecular Weight (g/mol) 174.2
Synonym Glutamine, N-ethyl- (9CI, ACI), DL-Glutamine, N-ethyl- (ZCI), Glutamine, N-ethyl-, DL- (8CI), N-Ethylglutamine (ACI), 2-Amino-5-(ethylamino)-5-oxopentanoic acid, DL-Theanine, N-Ethyl-DL-glutamine
IUPAC Name 2-amino-5-(ethylamino)-5-oxopentanoic acid
SMILES CCNC(=O)CCC(N)C(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C7H14N2O3/c1-2-9-6(10)4-3-5(8)7(11)12/h5H,2-4,8H2,1H3,(H,9,10)(H,11,12)
Formula Weight 174.1004
Chemical Name or Material DL-Theanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.