missing translation for 'onlineSavingsMsg'
Learn More

dl-tartaric acid, TRC

Product Code. 30357486
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357486 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357486 Supplier Toronto Research Chemicals Supplier No. TRCD285896100MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,3-Dihydroxybutanedioic Acid
CAS 526-83-0
Molecular Formula C4 H6 O6
Quantity 100 mg
Synonym Butanedioic acid, 2,3-dihydroxy-, 2,3-Dihydroxybutanedioic acid
InChI Formula InChI=1S/C4H6O6/c5-1(3(7)8)2(6)4(9)10/h1-2,5-6H,(H,7,8)(H,9,10)
SMILES OC(C(O)C(=O)O)C(=O)O
IUPAC Name 2,3-dihydroxybutanedioic acid
Molecular Weight (g/mol) 150.087
Formula Weight 150.016
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.