missing translation for 'onlineSavingsMsg'
Learn More

DL-Tartaric acid, 99%

Product Code. 11413454
Change view
Click to view available options
Quantity:
250 g
500 g
1000 g
Unit Size:
1000g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413454 1000 g 1000g
11423454 250 g 250g
11498950 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11413454 Supplier Thermo Scientific Alfa Aesar Supplier No. A10683.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 133-37-9 | C4H6O6 | 150.09 g/mol

DL-Tartaric acid is used as a synergist for antioxidants, emulsifier, sequestrant and flavoring agent. It is also added with citric acid to prepare effervescent salts, thereby enhancing the taste of oral medications. It is also utilized in pigments, processing aids, ink, toner and colorant products. It acts as a chelating agent in metal and farming industries. Further, it is used as lubricant and grease. It is mixed with sodium bicarbonate and used as a leavening agent in food preparation. In the pharmaceutical industry, it is utilized in the preparation of tartar emetic, which is used in cough syrup as an expectorant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Tartaric acid is used as a synergist for antioxidants, emulsifier, sequestrant and flavoring agent. It is also added with citric acid to prepare effervescent salts, thereby enhancing the taste of oral medications. It is also utilized in pigments, processing aids, ink, toner and colorant products. It acts as a chelating agent in metal and farming industries. Further, it is used as lubricant and grease. It is mixed with sodium bicarbonate and used as a leavening agent in food preparation. In the pharmaceutical industry, it is utilized in the preparation of tartar emetic, which is used in cough syrup as an expectorant.

Solubility
Soluble in water, alcohol, mineral acids and alkalies.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133-37-9
Molecular Formula C4H6O6
Molecular Weight (g/mol) 150.09
MDL Number MFCD00071626
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
Synonym dl-tartaric acid, 2,3-dihydroxysuccinic acid, tartaric acid, racemic acid, uvic acid, traubensaure, paratartaric acid, paratartaric aicd, dl-tartrate, racemic tartaric acid
PubChem CID 875
ChEBI CHEBI:15674
IUPAC Name 2,3-dihydroxybutanedioic acid
SMILES OC(C(O)C(O)=O)C(O)=O

Specifications

Melting Point 210°C to 212°C (decomposition)
Beilstein 1725148
Merck Index 14,9069
Solubility Information Soluble in water,alcohol,mineral acids and alkalies.
Formula Weight 150.09
Percent Purity 99%
Quantity 1000 g
Chemical Name or Material DL-Tartaric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.