missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Serine methyleester hydrochloride, 99%

Catalog Number 10178033
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
C4H9NO3·HCl
MFCD00012593
This item is not returnable. View return policy
C4H9NO3·HCl
MFCD00012593

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Assay Percent Range 99%
Infrared Spectrum Authentic
Linear Formula HOCH2CH(NH2)CO2CH3·HCl
Beilstein 04, 512
Packaging Glass Bottle
Formula Weight 155.58
Percent Purity ≥98.5%
Quantity 100 g
Physical Form Crystalline Powder
Chemical Name or Material DL-Serine methyleester hydrochloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'