missing translation for 'onlineSavingsMsg'
Learn More

DL-Serine, MedChemExpress

Product Code. 16449416
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16449416 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16449416 Supplier Medchem Express Supplier No. HYY0507
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress DL-Serine, a fundamental metabolite, is a mixture of D-Serine and L-Serine. DL-Serine has antiviral activity against the multiplication of tobacco mosaic virus (TMV).

DL-Serine, a fundamental metabolite, is a mixture of D-Serine and L-Serine. DL-Serine has antiviral activity against the multiplication of tobacco mosaic virus (TMV).

Specifications

Chemical Name or Material DL-Serine
Physical Form Solid
CAS 302-84-1
Color White to Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C3H7NO3
For Use With (Application) Cancer-programmed cell death
Solubility Information H2O : 33.33 mg/mL (317.16 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble or slightly soluble)
SMILES NC(CO)C(O)=O
Molecular Weight (g/mol) 105.09
Formula Weight 105.09
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.