missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Selenomethionine, 99+%

Product Code. 10410131 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
1 g
2.5 g
Packaging:
Glass bottle
Unit Size:
1g
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10410131 1 g Glass bottle 1g
10657592 500 mg Glass bottle 500mg
10450511 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10410131 Supplier Thermo Scientific Chemicals Supplier No. 240770010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1464-42-2 | C5H11NO2Se | 196.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1464-42-2
Molecular Formula C5H11NO2Se
Molecular Weight (g/mol) 196.12
MDL Number MFCD00063089
InChI Key RJFAYQIBOAGBLC-UHFFFAOYNA-N
Synonym selenomethionine, dl-selenomethionine, seleno-dl-methionine, +--selenomethionine, butanoic acid, 2-amino-4-methylseleno, selenium methionine, methionine, seleno, selenomethionine van, 2-amino-4-methylselanyl butanoic acid, seleno-d,l-methionine
PubChem CID 15103
ChEBI CHEBI:27585
IUPAC Name 2-amino-4-methylselanylbutanoic acid
SMILES C[Se]CCC(N)C(O)=O

Specifications

Melting Point 265.0°C
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Odor Faint Smelling
Packaging Glass bottle
Linear Formula CH3SeCH2CH2CH(NH2)COOH
Quantity 1 g
Merck Index 15, 8578
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol, insoluble in benzene, diethyl ether
Formula Weight 196.11
Percent Purity 99+%
Physical Form Glistening Powder
Chemical Name or Material DL-Selenomethionine, 99+%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • DL-Selenomethionine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.