missing translation for 'onlineSavingsMsg'
Learn More

DL-Pyroglutamic Acid, MP Biomedicals™

Product Code. 11496142 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g

Product Code. 11496142

Brand: MP Biomedicals 0210278080

Please to purchase this item. Need a web account? Register with us today!

Chemical Identifiers

CAS 149-87-1
Molecular Formula C5H7NO3
Molecular Weight (g/mol) 129.12
MDL Number MFCD00064322
InChI Key ODHCTXKNWHHXJC-UHFFFAOYNA-N
Synonym dl-pyroglutamic acid, 2-pyrrolidone-5-carboxylic acid, dl-proline, 5-oxo, 5-oxo-dl-proline, pyroglutamate, h-dl-pyr-oh, pyrrolidonecarboxylic acid, 5-oxoprolinate, d-+-pyroglutamic acid, dl-pidolic acid
PubChem CID 499
ChEBI CHEBI:16010
IUPAC Name 5-oxopyrrolidine-2-carboxylic acid
SMILES OC(=O)C1CCC(=O)N1

Specifications

Quantity 100 g
Formula Weight 129.1
Physical Form Solid
Chemical Name or Material DL-Pyroglutamic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.