missing translation for 'onlineSavingsMsg'
Learn More

DL-Propylhexedrine Hydrochloride, TRC

Product Code. 30012809
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012809 50 mg 50mg
30010055 10 mg 10mg
30005564 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012809 Supplier Toronto Research Chemicals Supplier No. TRCP83715050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1007-33-6
Molecular Formula C10 H21 N . Cl H
Molecular Weight (g/mol) 191.74
Synonym N,α-Dimethyl-cyclohexaneethanamine hydrochloride, N,α-dimethyl-cyclohexaneethylamine hydrochloride, (±)-Propylhexedrine hydrochloride, Benzedrex hydrochloride, Cyclohexyl(isopropyl)methylammonium chloride, Cyclohexylisopropylmethylamine hydrochloride, Eventin hydrochloride, NSC 170998, NSC 27110, Propylhexedrine hydrochloride, dl-Propylhexedrine hydrochloride
IUPAC Name 1-cyclohexyl-N-methylpropan-2-amine;hydrochloride
SMILES Cl.CNC(C)CC1CCCCC1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H21N.ClH/c1-9(11-2)8-10-6-4-3-5-7-10;/h9-11H,3-8H2,1-2H3;1H
Formula Weight 191.1441
Percent Purity >95%
Chemical Name or Material Propylhexedrine (hydrochloride)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.