missing translation for 'onlineSavingsMsg'
Learn More

DL-Phenylalanine, TRC

Product Code. 30411256
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30411256 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30411256 Supplier Toronto Research Chemicals Supplier No. TRCP1812132.5G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DL-Phenylalanine
CAS 150-30-1
Molecular Formula C9 H11 N O2
Quantity 2.5 g
Synonym Phenylalanine, Aspartame Imp. C (EP) as racemate, Alanine','phenyl-','DL- (8CI), Alanine','β-phenyl- (3CI), (±)-3-Phenyl-2-aminopropanoic acid, (±)-Phenylalanine, (±)-β-Phenyl-α-alanine, 2-Amino-3-phenylpropanoic acid, DL-2-Amino-3-phenylpropanoic acid, DL-2-Amino-3-phenylpropanoic acid, DL-2-Amino-3-phenylpropionic acid, DL-2-Amino-3-phenylpropionic acid, DL-3-Phenylalanine, DL-3-Phenylalanine, DL-Phenylalanine, DL-Phenylalanine, DL-beta-Phenyl-alpha-alanine, DL-α-Amino-β-phenylpropionic acid, DL-β-Phenyl-α-alanine, DLPA, NSC 9959
InChI Formula InChI=1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)
SMILES NC(Cc1ccccc1)C(=O)O
IUPAC Name 2-amino-3-phenylpropanoic acid
Molecular Weight (g/mol) 165.19
Formula Weight 165.079
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.