missing translation for 'onlineSavingsMsg'
Learn More

DL-Phenylalanine, ≥99%, MP Biomedicals™

Product Code. 11448250 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448250 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11448250 Supplier MP Biomedicals Supplier No. 0210261891
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 150-30-1
Color White
Assay Percent Range ≥99%
Molecular Formula C9H11NO2
MDL Number MFCD00064225
Synonym dl-phenylalanine, phenylalanine, h-dl-phe-oh, dl-3-phenylalanine, phenylalanin, dlpa, dl-2-amino-3-phenylpropanoic acid, alanine, phenyl-, dl, 2-amino-3-phenylpropionic acid, phenylalanine, dl
InChI Key COLNVLDHVKWLRT-UHFFFAOYNA-N
SMILES NC(CC1=CC=CC=C1)C(O)=O
IUPAC Name 2-amino-3-phenylpropanoic acid
Molecular Weight (g/mol) 165.19
Quantity 1 kg
PubChem CID 994
ChEBI CHEBI:28044
Formula Weight 165.2
Physical Form Powder
Chemical Name or Material DL-Phenylalanine, FTIR
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.