missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Phenylalanine, 99%

Product Code. 10357340 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10357340 100 g Plastic bottle 100g
10793541 25 g Plastic bottle 25g
11915341 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10357340 Supplier Thermo Scientific Chemicals Supplier No. 164571000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

DL-Phenylalanine, 99%, C9H11NO2, CAS Number-150-30-1, 2-amino-3-phenylpropionic acid, alanine, phenyl-, dl, dl-2-amino-3-phenylpropanoic acid, dl-3-phenylalanine, dlpa, dl-phenylalanine, h-dl-phe-oh, phenylalanin, phenylalanine, phenylalanine, dl, 100g, 14,498, CHEBI:28044, White, 205-756-7, 165.19 | CAS: 150-30-1 | C9H11NO2 | 165.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-30-1
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00064225
InChI Key COLNVLDHVKWLRT-UHFFFAOYNA-N
Synonym dl-phenylalanine, phenylalanine, h-dl-phe-oh, dl-3-phenylalanine, phenylalanin, dlpa, dl-2-amino-3-phenylpropanoic acid, alanine, phenyl-, dl, 2-amino-3-phenylpropionic acid, phenylalanine, dl
PubChem CID 994
ChEBI CHEBI:28044
IUPAC Name 2-amino-3-phenylpropanoic acid
SMILES NC(CC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 266.0°C
Color White
Loss on Drying 0.3% (105°C, 2 hrs) max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula C6H5CH2CH(NH2)CO2H
Quantity 100 g
Beilstein 14,498
Merck Index 15,7382
Solubility Information Solubility in water: 14.11g/L (25°C)
Formula Weight 165.19
Percent Purity 99%
Physical Form Powder
Chemical Name or Material DL-Phenylalanine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.