missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Phenylalanine, 99%

Product Code. 11419392
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419392 500 g 500g
11499382 25 g 25g
11409392 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11419392 Supplier Thermo Scientific Chemicals Supplier No. A10132.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 150-30-1 | C9H11NO2 | 165.19 g/mol

DL-Phenylalanine is used to relieve cases of minor depression and other mood issues as well as and physical discomforts and pain. It is also used in acupuncture anesthesia, alcohol withdrawal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Phenylalanine is used to relieve cases of minor depression and other mood issues as well as and physical discomforts and pain. It is also used in acupuncture anesthesia, alcohol withdrawal.

Solubility
Soluble in water. Insoluble in ethyl ether, benzene.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-30-1
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00064225
InChI Key COLNVLDHVKWLRT-UHFFFAOYNA-N
Synonym dl-phenylalanine, phenylalanine, h-dl-phe-oh, dl-3-phenylalanine, phenylalanin, dlpa, dl-2-amino-3-phenylpropanoic acid, alanine, phenyl-, dl, 2-amino-3-phenylpropionic acid, phenylalanine, dl
PubChem CID 994
ChEBI CHEBI:28044
IUPAC Name 2-amino-3-phenylpropanoic acid
SMILES NC(CC1=CC=CC=C1)C(O)=O

Specifications

Melting Point ∼266°C (decomposition)
Beilstein 1910407
Merck Index 14,7271
Solubility Information Soluble in water. Insoluble in ethyl ether,benzene.
Quantity 500 g
Formula Weight 165.19
Percent Purity 99%
Chemical Name or Material DL-Phenylalanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.