missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Phenylalanine, 99%

Catalog Number p-4510421
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
150-30-1
C9H11NO2
165.19
MFCD00064225
COLNVLDHVKWLRT-UHFFFAOYNA-N
dl-phenylalanine, phenylalanine, h-dl-phe-oh, dl-3-phenylalanine, phenylalanin, dlpa, dl-2-amino-3-phenylpropanoic acid, alanine, phenyl-, dl, 2-amino-3-phenylpropionic acid, phenylalanine, dl
994
CHEBI:28044
2-amino-3-phenylpropanoic acid
NC(CC1=CC=CC=C1)C(O)=O
This item is not returnable. View return policy
150-30-1
C9H11NO2
165.19
MFCD00064225
COLNVLDHVKWLRT-UHFFFAOYNA-N
dl-phenylalanine, phenylalanine, h-dl-phe-oh, dl-3-phenylalanine, phenylalanin, dlpa, dl-2-amino-3-phenylpropanoic acid, alanine, phenyl-, dl, 2-amino-3-phenylpropionic acid, phenylalanine, dl
994
CHEBI:28044
2-amino-3-phenylpropanoic acid
NC(CC1=CC=CC=C1)C(O)=O

DL-Phenylalanine, 99%, C9H11NO2, CAS Number-150-30-1, 2-amino-3-phenylpropionic acid, alanine, phenyl-, dl, dl-2-amino-3-phenylpropanoic acid, dl-3-phenylalanine, dlpa, dl-phenylalanine, h-dl-phe-oh, phenylalanin, phenylalanine, phenylalanine, dl, 100g, 14,498, CHEBI:28044, White, 205-756-7, 165.19

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 266.0°C
Color White
Loss on Drying 0.3% (105°C, 2 hrs) max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula C6H5CH2CH(NH2)CO2H
Quantity 25 g
Beilstein 14,498
Merck Index 15,7382
Solubility Information Solubility in water: 14.11g/L (25°C)
Formula Weight 165.19
Percent Purity 99%
Physical Form Powder
Chemical Name or Material DL-Phenylalanine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'