missing translation for 'onlineSavingsMsg'
Learn More

Dl-Phenylalanine, 98.5+%, Extra Pure, Fisher Chemical

Product Code. 10365950 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10365950 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10365950 Supplier Fisher Chemical Supplier No. P/2461/48

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 266°C
CAS 150-30-1
Color White
Molecular Formula C9H11NO2
Packaging Plastic Powder Jar
MDL Number MFCD00064225
Synonym dl-phenylalanine, phenylalanine, h-dl-phe-oh, dl-3-phenylalanine, phenylalanin, dlpa, dl-2-amino-3-phenylpropanoic acid, alanine, phenyl-, dl, 2-amino-3-phenylpropionic acid, phenylalanine, dl
InChI Key COLNVLDHVKWLRT-UHFFFAOYNA-N
SMILES NC(CC1=CC=CC=C1)C(O)=O
IUPAC Name 2-amino-3-phenylpropanoic acid
Molecular Weight (g/mol) 165.19
Quantity 100 g
PubChem CID 994
ChEBI CHEBI:28044
Formula Weight 165.19g/mol
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.