missing translation for 'onlineSavingsMsg'
Learn More

DL-Penicillamine, 97+%

Product Code. 11450598
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11450598

Brand: Thermo Scientific Alfa Aesar B24710.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 52-66-4 | C5H11NO2S | 149.21 g/mol

Heavy metal chelating agentDL-Penicillamine is used as an antirheumatic. It also serves as a chelating agent for heavy metals. Further, it is used in the treatment of Wilson's desease, Cystinuria, Scleroderma and arsenic poisoning.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Heavy metal chelating agentDL-Penicillamine is used as an antirheumatic. It also serves as a chelating agent for heavy metals. Further, it is used in the treatment of Wilson′s desease, Cystinuria, Scleroderma and arsenic poisoning.

Solubility
Soluble in ethanol, hot water and sodium hydroxide.

Notes
Incompatible with oxidizing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52-66-4
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.21
MDL Number MFCD00004856
InChI Key VVNCNSJFMMFHPL-UHFFFAOYNA-N
Synonym dl-penicillamine, 2-amino-3-mercapto-3-methylbutanoic acid, +--penicillamine, 3,3-dimethyl-dl-cysteine, 3-sulfanylvaline, dl-beta-mercaptovaline, beta-mercaptovaline, d,l-penicillamine, dl-3-mercaptovaline, 2-amino-3-mercapto-3-methylbutyric acid
PubChem CID 4727
ChEBI CHEBI:50868
IUPAC Name 2-amino-3-methyl-3-sulfanylbutanoic acid
SMILES CC(C)(S)C(N)C(O)=O

Specifications

Melting Point ∼212°C (decomposition)
Odor Unpleasant
Quantity 5 g
Beilstein 2039817
Merck Index 14,7088
Solubility Information Soluble in ethanol,hot water and sodium hydroxide.
Formula Weight 149.21
Percent Purity ≥97%
Chemical Name or Material DL-Penicillamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.