missing translation for 'onlineSavingsMsg'
Learn More

DL-Panthenol, MedChemExpress

Product Code. 16482299
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482299 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16482299 Supplier Medchem Express Supplier No. HYB1024
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress DL-Panthenol (DL-Pantothenol) is an alcohol derivative of pantothenyl acid. DL-Panthenol exerts eyelash protection effect. DL-Panthenol is widely used in the Skin and hair conditioner research.

DL-Panthenol (DL-Pantothenol) is an alcohol derivative of pantothenyl acid. DL-Panthenol exerts eyelash protection effect. DL-Panthenol is widely used in the Skin and hair conditioner research.

Specifications

Chemical Name or Material DL-Panthenol
CAS 16485-10-2
Color White
Purity Grade Notes Research
Molecular Formula C9H19NO4
Quantity 1 g
Synonym DL-Pantothenol DL-Pantothenyl alcohol
Solubility Information DMSO : ≥ 46 mg/mL (224.12 mM)
SMILES O=C(NCCCO)C(O)C(C)(C)CO
Molecular Weight (g/mol) 205.25
Formula Weight 205.25
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.