missing translation for 'onlineSavingsMsg'
Learn More

DL-p-Hydroxyphenyllactic Acid-d3, TRC

Product Code. 30395752
Change view
Click to view available options
Quantity:
1 mg
10 mg
100 mg
Unit Size:
100mg
10mg
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395752 1 mg 1mg
30403302 10 mg 10mg
30401978 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30395752 Supplier Toronto Research Chemicals Supplier No. TRCH9536671MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DL-p-Hydroxyphenyllactic Acid-d3
Purity Grade Notes HPLC
Molecular Formula C9H7D3O4
Quantity 1 mg
Synonym 3-(p-Hydroxyphenyl)lactic Acid, (RS)-3-(4-Hydroxyphenyl)lactic Acid-d3, (±)-3-(4-Hydroxyphenyl)lactic Acid-d3, 2-Hydroxy-3-(4-hydroxyphenyl)propanoic Acid-d3, 2-Hydroxy-3-(p-hydroxyphenyl)propionic Acid-d3, 3-(4-Hydroxyphenyl)-DL-lactic Acid-d3, 3-(4-Hydroxyphenyl)lactic Acid-d3, DL-3-(4-Hydroxyphenyl)lactic Acid-d3, HPLA-d3, NSC 111175-d3, p-Hydroxy-dl-phenyllactic Acid-d3, α,4-Dihydroxybenzenepropanoic Acid-d3, β-(4-Hydroxyphenyl)lactic Acid-d3, β-(p-Hydroxyphenyl)-DL-lactic Acid-d3, β-(p-Hydroxyphenyl)lactic Acid-d3
InChI Formula InChI=1S/C9H10O4/c10-7-3-1-6(2-4-7)5-8(11)9(12)13/h1-4,8,10-11H,5H2,(H,12,13)/i5D2,8D
SMILES O=C(O)C([2H])(O)C([2H])([2H])C1=CC=C(O)C=C1
Molecular Weight (g/mol) 185.19
Percent Purity >95
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.