missing translation for 'onlineSavingsMsg'
Learn More

DL-Norvaline, MedChemExpress

Product Code. 16462678
Encompass_Preferred
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 16462678

Brand: Medchem Express HYW010510

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress DL-Norvaline, a derivative of L-norvaline, L-norvaline is a non-competitive inhibitor of arginase.

DL-Norvaline, a derivative of L-norvaline, L-norvaline is a non-competitive inhibitor of arginase.

Specifications

Chemical Name or Material DL-Norvaline
CAS 760-78-1
Color White
Purity Grade Notes Research
Molecular Formula C5H11NO2
Synonym 2-Aminopentanoic acid
Solubility Information H2O : 50 mg/mL (426.80 mM; Need ultrasonic)
SMILES CCCC(N)C(O)=O
Molecular Weight (g/mol) 117.15
Quantity 5 g
Formula Weight 117.15
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.