missing translation for 'onlineSavingsMsg'
Learn More

DL-Nornicotine, 96%

Product Code. 11363006
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11363006 50 mg 50mg
11353006 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11363006 Supplier Thermo Scientific Alfa Aesar Supplier No. L10809.MF

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5746-86-1 | C9H12N2 | 148.209 g/mol

DL-Nornicotine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Nornicotine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in acetone, chloroform and water (50mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature: 2 - 8°C. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5746-86-1
Molecular Formula C9H12N2
Molecular Weight (g/mol) 148.209
MDL Number MFCD00016913
InChI Key MYKUKUCHPMASKF-UHFFFAOYSA-N
Synonym 3-pyrrolidin-2-yl pyridine, nornicotine, 3-2-pyrrolidinyl pyridine, r,s-nornicotine, +--nornicotine, 2-3-pyridyl pyrrolidine, 3-pyrrolidin-2-yl-pyridine, +--1'-demethylnicotine, +/--nornicotine, dl-nornicotine
PubChem CID 412
IUPAC Name 3-pyrrolidin-2-ylpyridine
SMILES C1CC(NC1)C2=CN=CC=C2

Specifications

Density 1.074
Boiling Point 107°C to 108°C (2 mmHg)
Flash Point 101°C (213°F)
Refractive Index 1.549
Quantity 50 mg
UN Number UN2810
Beilstein 81968
Sensitivity Hygroscopic
Merck Index 14,6712
Solubility Information Soluble in acetone,chloroform and water (50mg/ml).
Formula Weight 148.21
Percent Purity 98%
Chemical Name or Material DL-Nornicotine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.