missing translation for 'onlineSavingsMsg'
Learn More

DL-N-Propyl Sulfite, TRC

Product Code. 30380055
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380055 2.5 g 2.5g
30378615 500 mg -
30370006 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380055 Supplier Toronto Research Chemicals Supplier No. TRCD6799402.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-98-3
Molecular Formula C6 H14 O3 S
Molecular Weight (g/mol) 166.239
IUPAC Name dipropyl sulfite
SMILES CCCOS(=O)OCCC

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C6H14O3S/c1-3-5-8-10(7)9-6-4-2/h3-6H2,1-2H3
Formula Weight 166.066
Chemical Name or Material DL-N-Propyl Sulfite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.