missing translation for 'onlineSavingsMsg'
Learn More

DL-alpha-Methylbenzylamine, 99%

Product Code. p-159806 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10295410

Brand: Thermo Scientific Acros 126390050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

DL-alpha-Methylbenzylamine, 99%, C8H11N, CAS Number-618-36-0, 1-amino-1-phenylethane, 1-phenethylamine, 1-phenylethan-1-amine, 1-phenylethylamine, alpha-aminoethylbenzene, alpha-methylbenzenemethanamine, alpha-methylbenzylamine, alpha-phenylethylamine, a-methylbenzylamine, dl-alpha-methylbenzylamine | CAS: 618-36-0 | C8H11N | 121.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 618-36-0
MDL Number MFCD00008069
InChI Key RQEUFEKYXDPUSK-UHFFFAOYSA-N
Synonym 1-phenylethylamine, alpha-methylbenzylamine, alpha-phenylethylamine, dl-alpha-methylbenzylamine, 1-phenethylamine, alpha-aminoethylbenzene, 1-amino-1-phenylethane, 1-phenylethan-1-amine, a-methylbenzylamine, alpha-methylbenzenemethanamine
PubChem CID 7408
ChEBI CHEBI:670
IUPAC Name 1-phenylethanamine
SMILES CC(C1=CC=CC=C1)N

Specifications

Color Colorless to Yellow
Density 0.94g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C8H11N
Linear Formula C6H5CH(CH3)NH2
Refractive Index 1.5250 to 1.5270 (20°C, 589nm)
Quantity 5 g
Beilstein 12, 1094
Merck Index 14, 6026
Molecular Weight (g/mol) 121.183
Formula Weight 121.18
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material DL-α-Methylbenzylamine, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.