missing translation for 'onlineSavingsMsg'
Learn More

DL-Mercaptosuccinic acid, 99%

Product Code. 10133270 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10133270 500 g Glass bottle 500g
10574051 100 g Glass bottle 100g
11995721 2.5 kg Plastic Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10133270 Supplier Thermo Scientific Acros Supplier No. 125555000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 70-49-5 | C4H6O4S | 150.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70-49-5
Molecular Formula C4H6O4S
Molecular Weight (g/mol) 150.15
InChI Key NJRXVEJTAYWCQJ-UHFFFAOYSA-N
Synonym mercaptosuccinic acid, thiomalic acid, 2-thiomalic acid, 2-mercaptosuccinic acid, monomercaptosuccinic acid, butanedioic acid, mercapto, malic acid, 2-thio, mercaptosuccinate, 2-sulfanylsuccinic acid, dl-mercaptosuccinic acid
PubChem CID 6268
ChEBI CHEBI:38705
IUPAC Name 2-sulfanylbutanedioic acid
SMILES C(C(C(=O)O)S)C(=O)O

Specifications

Melting Point 152°C to 155°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula HO2CCH2CH(SH)CO2H
Quantity 500 g
Beilstein 03,439
Merck Index 15,9496
Solubility Information Solubility in water: 150g/L (20°C). Other solubilities: 500g/L ethanol (20°C),soluble in acetone,moderately soluble in ether,chloroform,practically insoluble in benzene
Formula Weight 150.15
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material DL-Mercaptosuccinic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.