missing translation for 'onlineSavingsMsg'
Learn More

DL-Malic Acid, TRC

Product Code. 30011757
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011757 100 g 100g
30009962 10 g 10g
17970665 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011757 Supplier Toronto Research Chemicals Supplier No. TRCM159510100G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6915-15-7
Molecular Formula C4 H6 O5
Molecular Weight (g/mol) 134.09
Synonym Malic acid, (2RS)-2-Hydroxybutanedioic acid, Butanedioic acid, hydroxy- (9CI), Malic acid (8CI), (±)-Malic acid, 2-Hydroxybutanedioic acid, 2-Hydroxyethane-1,2-dicarboxylic acid, 2-Hydroxysuccinic acid, DL-Malic acid, Deoxytetraric acid, E 296, FDA 2018, Hydroxybutanedioic acid, Hydroxysuccinic acid, Musashi-no-Ringosan, NSC 25941, Nanoveson M, Pomalus Acid, R,S(±)-Malic acid, Xeros, dl-Malic acid, α-Hydroxysuccinic acid
IUPAC Name 2-hydroxybutanedioic acid
SMILES OC(CC(=O)O)C(=O)O

Specifications

Quantity 100 g
InChI Formula InChI=1S/C4H6O5/c5-2(4(8)9)1-3(6)7/h2,5H,1H2,(H,6,7)(H,8,9)
Formula Weight 134.0215
Percent Purity >95%
Chemical Name or Material DL-Malic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.