missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Malic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
2500 g
Description
DL-Malic acid is used as a food additive. It is a component of some artificial vinegar flavors. Further, it is used in the preparation of chiral compounds, including κ-opioid rece. It is utilized for chiral resolution by ligand-exchange capillary electrophoresis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
DL-Malic acid is used as a food additive. It is a component of some artificial vinegar flavors. Further, it is used in the preparation of chiral compounds, including κ-opioid rece. It is utilized for chiral resolution by ligand-exchange capillary electrophoresis.
Solubility
Soluble in acetone, ether, water, methanol and ethanol.
Notes
Incompatible with bases, oxidizing agents, reducing agents and alkali metals.
DL-Malic acid is used as a food additive. It is a component of some artificial vinegar flavors. Further, it is used in the preparation of chiral compounds, including κ-opioid rece. It is utilized for chiral resolution by ligand-exchange capillary electrophoresis.
Solubility
Soluble in acetone, ether, water, methanol and ethanol.
Notes
Incompatible with bases, oxidizing agents, reducing agents and alkali metals.
Specifications
Melting Point | 130°C to 133°C |
Assay Percent Range | 0.98 |
Beilstein | 1723539 |
Solubility Information | Soluble in acetone,ether,water,methanol and ethanol. |
Formula Weight | 134.09 |
Density | 1.609 |
Quantity | 500 g |
Flash Point | 203°C (397°F) |
Chemical Name or Material | DL-Malic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'