missing translation for 'onlineSavingsMsg'
Learn More

DL-Lysine monohydrochloride, 99%

Product Code. 10289740 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass Bottle
Unit Size:
100g
1kg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10289740 100 g Glass Bottle 100g
10710102 25 g Glass Bottle 25g
10300684 1 kg Glass Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10289740 Supplier Thermo Scientific Acros Supplier No. 413371000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 70-53-1 | C6H15ClN2O2 | 182.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70-53-1
Molecular Formula C6H15ClN2O2
Molecular Weight (g/mol) 182.65
MDL Number MFCD00064563
InChI Key BVHLGVCQOALMSV-UHFFFAOYSA-N
Synonym dl-lysine monohydrochloride, 2,6-diaminohexanoic acid hydrochloride, dl-lysine hydrochloride, lysine monohydrochloride, dl-lysine mono hydrochloride, lysine, hydrochloride 1:1, lysine, monohydrochloride, dl-2,6-diaminohexanoic acid monohydrochloride, lysine, dl, dl-lysinemonohydrochloride
PubChem CID 12047
IUPAC Name 2,6-diaminohexanoic acid;hydrochloride
SMILES Cl.NCCCCC(N)C(O)=O

Specifications

Melting Point 267.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula NH2(CH2)4CH(NH2)COO·HCl
Specific Rotation −1° to +1° (20°C, 589nm) (c=1, water)
Merck Index 14, 5636
Quantity 100 g
Formula Weight 182.65
Percent Purity 99%
Physical Form Powder, Granules or Chunks
Chemical Name or Material DL-Lysine monohydrochloride, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.