missing translation for 'onlineSavingsMsg'
Learn More

DL-leucine, TRC

Product Code. 17969924
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17969924 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17969924 Supplier Toronto Research Chemicals Supplier No. TRCL330170500MG
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 328-39-2
Molecular Formula C6 H13 N O2
Synonym Leucine, DL-Leucine, Leucine, DL- (8CI), (RS)-Leucine, (±)-Leucine, 2-Amino-4-methylpentanoic acid, NSC 9252
InChI Formula InChI=1S/C6H13NO2/c1-4(2)3-5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)
SMILES CC(C)CC(N)C(=O)O
IUPAC Name 2-amino-4-methylpentanoic acid
Molecular Weight (g/mol) 131.17
Quantity 500 mg
Formula Weight 131.0946
Shipping Condition Room Temperature
Chemical Name or Material DL-Leucine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.