missing translation for 'onlineSavingsMsg'
Learn More

DL-Lactic acid, 90%

Product Code. 10487510 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
10kg
1kg
2.5kg
25g
500g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10487510 2.5 kg Glass bottle 2.5kg
10736341 25 g Glass bottle 25g
10153460 500 g Glass bottle 500g
10031120 1 kg Glass bottle 1kg
10554074 10 kg Plastic drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10487510 Supplier Thermo Scientific Acros Supplier No. 125060025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 50-21-5 | C3H6O3 | 90.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-21-5 , 7732-18-5
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00004520
InChI Key JVTAAEKCZFNVCJ-UHFFFAOYNA-N
Synonym lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum
PubChem CID 612
ChEBI CHEBI:78320
IUPAC Name 2-hydroxypropanoic acid
SMILES CC(O)C(O)=O

Specifications

Melting Point 18.0°C
Boiling Point 122.0°C (15.0 mmHg)
Color Colorless to Yellow
Linear Formula CH3CH(OH)CO2H
Merck Index 15, 5384
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, furfurol, glycerol, less soluble in ether, practically insoluble in chloroform, carbon, disulfide, petroleum ether
Formula Weight 90.08
Percent Purity 90%
Density 1.2000g/mL
Quantity 2.5 kg
Flash Point >110°C
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.4252 to 1.4272
Residue after Ignition 0.1% max.
Specific Gravity 1.2
Chemical Name or Material DL-Lactic acid
Show More Show Less
compliance-icons
Product Identifier
  • DL-Lactic acid
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 C
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.