missing translation for 'onlineSavingsMsg'
Learn More

DL-Isoserine, 98%

Product Code. 15430198
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15430198 5 g 5g
15420198 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15430198 Supplier Thermo Scientific Alfa Aesar Supplier No. H53412.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 565-71-9 | C3H7NO3 | 105.093 g/mol

DL-Isoserine has been used in the synthesis of L-isoserine derivatives s as aminopeptidase N inhibitors, Syntheses of (2R,3S)-3-Hydroxyleucinate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Isoserine has been used in the synthesis of L-isoserine derivatives s as aminopeptidase N inhibitors, Syntheses of (2R,3S)-3-Hydroxyleucinate.

Solubility
Soluble in HCl.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 565-71-9
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.093
MDL Number MFCD00008138
InChI Key BMYNFMYTOJXKLE-UHFFFAOYSA-N
Synonym dl-isoserine, isoserine, 3-amino-2-hydroxypropionic acid, propanoic acid, 3-amino-2-hydroxy, 2-hydroxy-3-aminopropanoic acid, h-dl-isoser-oh, inverted exclamation marka-3-amino-2-hydroxypropionic acid, rs-isoserine, pubchem23947
PubChem CID 11267
IUPAC Name 3-amino-2-hydroxypropanoic acid
SMILES C(C(C(=O)O)O)N

Specifications

Melting Point 235°C (decomposition)
Beilstein 1721413
Solubility Information Soluble in HCl.
Quantity 5 g
Formula Weight 105.09
Percent Purity 98%
Chemical Name or Material DL-Isoserine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.