Learn More
DL-Isoproterenol hydrochloride, 98%, Thermo Scientific™
Standard selective beta-adrenoceptor agonist
£108.65
Chemical Identifiers
| CAS | 51-30-9 |
|---|---|
| Molecular Formula | C11H18ClNO3 |
| Molecular Weight (g/mol) | 247.72 |
| MDL Number | MFCD00012603,MFCD00064548 |
| InChI Key | IROWCYIEJAOFOW-UHFFFAOYNA-N |
| Synonym | isoprenaline hydrochloride, isoproterenol hydrochloride, isuprel, euspiran, isoprenaline hcl, isoproterenol hcl, aerolone, aerotrol, izadrin, vapo-iso |
| PubChem CID | 5807 |
| IUPAC Name | hydrogen 4-{1-hydroxy-2-[(propan-2-yl)amino]ethyl}benzene-1,2-diol chloride |
| SMILES | [H+].[Cl-].CC(C)NCC(O)C1=CC=C(O)C(O)=C1 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIsoproterenol hydrochloride is used as a selective β-adrenergic agonist, increases cytosolic cAMP.
Solubility
Soluble in water (50 mg/ml at 25°C), ethanol (20 mg/ml), methanol, DMSO (50 mg/ml at 25°C), and phosphate buffered saline (50 mM).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Chemical Identifiers
| C11H18ClNO3 | |
| MFCD00012603,MFCD00064548 | |
| isoprenaline hydrochloride, isoproterenol hydrochloride, isuprel, euspiran, isoprenaline hcl, isoproterenol hcl, aerolone, aerotrol, izadrin, vapo-iso | |
| hydrogen 4-{1-hydroxy-2-[(propan-2-yl)amino]ethyl}benzene-1,2-diol chloride |
Specifications
| 51-30-9 | |
| MFCD00012603,MFCD00064548 | |
| 14,5217 | |
| Soluble in water (50mg/ml at 25°C),ethanol (20mg/ml),methanol,DMSO (50mg/ml at 25°C),and phosphate buffered saline (50 mM). | |
| [H+].[Cl-].CC(C)NCC(O)C1=CC=C(O)C(O)=C1 | |
| 247.72 | |
| 247.72 | |
| DL-Isoproterenol hydrochloride |
| C11H18ClNO3 | |
| 3917363 | |
| isoprenaline hydrochloride, isoproterenol hydrochloride, isuprel, euspiran, isoprenaline hcl, isoproterenol hcl, aerolone, aerotrol, izadrin, vapo-iso | |
| IROWCYIEJAOFOW-UHFFFAOYNA-N | |
| hydrogen 4-{1-hydroxy-2-[(propan-2-yl)amino]ethyl}benzene-1,2-diol chloride | |
| 5807 | |
| 98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 200-089-8
missing translation for 'rtecsNumber' : DO1925000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.