missing translation for 'onlineSavingsMsg'
Learn More

DL-Isoleucine, 99%

Product Code. 11448960
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
Unit Size:
100g
10g
25g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11448960 100 g 100g
11478137 10 g 10g
11438960 25 g 25g
11488137 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11448960 Supplier Thermo Scientific Alfa Aesar Supplier No. A17521.22
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 443-79-8 | C6H13NO2 | 131.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 443-79-8
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00004268
InChI Key AGPKZVBTJJNPAG-WHFBIAKZSA-N
PubChem CID 94206
ChEBI CHEBI:20899
SMILES CC[C@H](C)[C@H](N)C(O)=O

Specifications

Melting Point ∼280°C (decomposition)
Beilstein 1721796
Merck Index 14,5179
Synonym d-alloisoleucine, d-allo-isoleucine, 2r,3s-2-amino-3-methylpentanoic acid, allo-d-isoleucine, alloisoleucine, d, h-d-allo-ile-oh, threo-d-isoleucine, dl-allo-isoleucine, hile, h-dl-allo-ile-oh
IUPAC Name (2R,3S)-2-amino-3-methylpentanoic acid
Quantity 100 g
Formula Weight 131.18
Percent Purity 99%
Chemical Name or Material DL-Isoleucine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.