missing translation for 'onlineSavingsMsg'
Learn More

DL-Isocitric acid, trisodium salt hydrate, 95%

Product Code. 10053573 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10053573 25 g Glass bottle 25g
10388050 1 g Glass bottle 1g
10311054 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10053573 Supplier Thermo Scientific Acros Supplier No. 205010250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1637-73-6 | C6H5Na3O7 | 258.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1637-73-6
Molecular Formula C6H5Na3O7
Molecular Weight (g/mol) 258.07
MDL Number MFCD00149284
InChI Key HWMVXEKEEAIYGB-UHFFFAOYNA-K
Synonym dl-isocitric acid, isocitric acid trisodium salt, sodium isocitrate, dl-isocitric acid trisodium salt hydrate, dl-isocitric acid trisodium salt, trisodium 1-hydroxypropane-1,2,3-tricarboxylate, trisodium isocitrate, tripotassium isocitrate, isocitric acid, dl, dl-isocitric acid, trisodium salt hydrate
PubChem CID 168942
IUPAC Name trisodium;1-hydroxypropane-1,2,3-tricarboxylate
SMILES [Na+].[Na+].[Na+].OC(C(CC([O-])=O)C([O-])=O)C([O-])=O

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula NaO2CCH2CH(CO2Na)CH(OH)CO2Na·xH2O
Quantity 25 g
Beilstein 03, 555
Solubility Information Solubility in water: soluble in water
Formula Weight 258.07
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material DL-Isocitric acid, trisodium salt hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.