missing translation for 'onlineSavingsMsg'
Learn More

DL-Isoborneol, 93%, tech.

Product Code. 10705011 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
Packaging:
Plastic bottle
Unit Size:
25g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10705011 500 g Plastic bottle 500g
10488270 25 g Plastic bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10705011 Supplier Thermo Scientific Acros Supplier No. 122495000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 124-76-5 | C10H18O | 154.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-76-5
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00074821
InChI Key DTGKSKDOIYIVQL-SZBHIRRCSA-N
Synonym dl-isoborneol
PubChem CID 126961757
IUPAC Name (1R,3R)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-ol
SMILES CC1(C2CCC1(C(C2)O)C)C

Specifications

Melting Point 208.0°C to 214.0°C
Color Yellow
Flash Point 74°C
Infrared Spectrum Authentic
Assay Percent Range 91% min. (GC)
Packaging Plastic bottle
Quantity 500 g
Beilstein 06, II, 80
Merck Index 15, 5173
Solubility Information Solubility in water: insoluble in water. Other solubilities: readily soluble in alcohol,chloroform and ether
Formula Weight 154.25
Percent Purity 93%
Grade Technical
Physical Form Powder
Chemical Name or Material DL-Isoborneol, tech., 93%
Show More Show Less
compliance-icons
Product Identifier
  • DL-Isoborneol
Signal Word
  • Warning
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.