missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Indole-3-lactic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | DL-Indole-3-lactic Acid |
| CAS | 1821-52-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C11H11NO3 |
| Quantity | 500 mg |
| Synonym | Indole-3-lactic Acid, Indolelactic Acid, 2-Hydroxy-3-(1H-indol-3-yl)propanoic Acid, 2-Hydroxy-3-(3-indolyl)propanoic Acid, 3-(3-Indolyl)-2-hydroxypropionic Acid, 3-(3-Indolyl)-DL-lactic Acid, 3-Indolyllactic Acid, α-Hydroxy-1H-indole-3-propanoic Acid, β-(3-Indolyl)lactic Acid, β-Indolyllactic Acid |
| InChI Formula | InChI=1S/C11H11NO3/c13-10(11(14)15)5-7-6-12-9-4-2-1-3-8(7)9/h1-4,6,10,12-13H,5H2,(H,14,15) |
| SMILES | OC(Cc1c[nH]c2ccccc12)C(=O)O |
| IUPAC Name | 2-hydroxy-3-(1H-indol-3-yl)propanoic acid |
| Molecular Weight (g/mol) | 205.21 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?