missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Indole-3-lactic Acid, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | DL-Indole-3-lactic Acid |
| CAS | 1821-52-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C11H11NO3 |
| Quantity | 1 g |
| Synonym | Indole-3-lactic Acid, Indolelactic Acid, 2-Hydroxy-3-(1H-indol-3-yl)propanoic Acid, 2-Hydroxy-3-(3-indolyl)propanoic Acid, 3-(3-Indolyl)-2-hydroxypropionic Acid, 3-(3-Indolyl)-DL-lactic Acid, 3-Indolyllactic Acid, α-Hydroxy-1H-indole-3-propanoic Acid, β-(3-Indolyl)lactic Acid, β-Indolyllactic Acid |
| InChI Formula | InChI=1S/C11H11NO3/c13-10(11(14)15)5-7-6-12-9-4-2-1-3-8(7)9/h1-4,6,10,12-13H,5H2,(H,14,15) |
| SMILES | OC(Cc1c[nH]c2ccccc12)C(=O)O |
| IUPAC Name | 2-hydroxy-3-(1H-indol-3-yl)propanoic acid |
| Molecular Weight (g/mol) | 205.21 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction